3,5-Difluorobenzenesulfonamide 97% (by HPLC)
Supplier: Ambeed
|
Warning
|
A566036-1G
A566036-25G
A566036-5G
77103-650EA
11
USD
77103-650
77103-652
77103-654
3,5-Difluorobenzenesulfonamide 97% (by HPLC)
3,5-Difluorobenzenesulphonamide
Precaución: for Research Use Only
Formula:
C₆H₅F₂NO₂S |
MDL Number:
MFCD02091379 CAS Number: 140480-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...