tert-Butyl 5-bromofuran-2-carboxylate 95+%
Supplier: Ambeed
|
Warning
|
A565799-1G
A565799-250MG
A565799-5G
77103-644EA
209
USD
77103-644
77103-646
77103-648
tert-Butyl 5-bromofuran-2-carboxylate 95+%
tert-Butyl 5-bromofuran-2-carboxylate
Precaución: for Research Use Only
Formula:
C₉H₁₁BrO₃ MW: 247.08 g/mol |
MDL Number:
MFCD06657484 CAS Number: 59862-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...