[3-(1H-Pyrazol-3-yl)phenyl]boronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(3-(1H-Pyrazol-3-yl)phenyl)boronic acid
A564366-100MG
A564366-1G
A564366-250MG
77103-638EA
65
USD
77103-638
77103-640
77103-642
[3-(1H-Pyrazol-3-yl)phenyl]boronic acid 98%
[3-(1H-Pyrazol-3-yl)phenyl]boronic acid
Precaución: for Research Use Only
Formula:
C₉H₉BN₂O₂ |
MDL Number:
MFCD09701957 CAS Number: 1100095-25-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...