Atorvastatin calcium salt trihydrate 95%

Supplier: Ambeed
Warning

Synonyms: Calcium (3R,5R)-7-(2-(4-fluorophenyl)-5-isopropyl-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl)-3,5-dihydroxyheptanoate trihydrate, Atorvastatin Calcium Salt Trihydrate, Atorvastatin hemicalcium salt sesquihydrate, Atorvastatin calcium

A563943-100G A563943-1G A563943-5G
77103-632EA 276.9 USD
77103-632 77103-634 77103-636
Atorvastatin calcium salt trihydrate 95%
Atorvastatin calcium salt trihydrate

Precaución: for Research Use Only

Formula: C₆₆H₇₄CaF₂N₄O₁₃
MW: 1209.39 g/mol
MDL Number: MFCD09752074
CAS Number: 344423-98-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR