4,6-Dibromo-3-fluoro-2-methylaniline 97+%

Supplier: Ambeed
Warning

A563686-10G A563686-1G A563686-250MG A563686-5G
77103-624EA 800.98 USD
77103-624 77103-626 77103-628 77103-630
4,6-Dibromo-3-fluoro-2-methylaniline 97+%
4,6-Dibromo-3-fluoro-2-methylaniline

Precaución: for Research Use Only

Formula: C₇H₆Br₂FN
MW: 282.93 g/mol
MDL Number: MFCD09800762
CAS Number: 1000576-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR