Boc-His(Trt)-OH 98%

Supplier: Ambeed
Warning

A563393-100G A563393-10G A563393-1G A563393-25G A563393-500G A563393-5G
77103-612EA 197.33 USD
77103-612 77103-614 77103-616 77103-618 77103-620 77103-622
Boc-His(Trt)-OH 98%
Boc-His(Trt)-OH

Precaución: for Research Use Only

Formula: C₃₀H₃₁N₃O₄
MW: 497.6 g/mol
MDL Number: MFCD00153307
CAS Number: 32926-43-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR