4-Fluoro-2-(methylthio)phenylboronic acid 98%

Supplier: Ambeed
Warning

A563200-100MG A563200-1G A563200-250MG
77103-606EA 48.81 USD
77103-606 77103-608 77103-610
4-Fluoro-2-(methylthio)phenylboronic acid 98%
4-Fluoro-2-(methylthio)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BFO₂S
MW: 186.01 g/mol
MDL Number: MFCD18087696
CAS Number: 861931-38-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR