3-Chloro-4-iodopyridin-2-amine 95%

Supplier: Ambeed
Warning

A562626-1G A562626-250MG A562626-5G
77103-600EA 11.67 USD
77103-600 77103-602 77103-604
3-Chloro-4-iodopyridin-2-amine 95%
3-Chloro-4-iodopyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄ClIN₂
MW: 254.45 g/mol
MDL Number: MFCD22627829
CAS Number: 1152617-24-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR