(2S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(adamantan-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Adamantyl-Gly-OH

A562623-1G A562623-250MG
77103-596EA 748 USD
77103-596 77103-598
(2S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(adamantan-1-yl)acetic acid 95%
(2S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-(adamantan-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₂₇H₂₉NO₄
MDL Number: MFCD09952624
CAS Number: 1221793-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR