Ethyl 5-(cyanomethyl)furan-2-carboxylate 95+%

Supplier: Ambeed
Warning

A561987-1G A561987-250MG
77103-592EA 51.99 USD
77103-592 77103-594
Ethyl 5-(cyanomethyl)furan-2-carboxylate 95+%
Ethyl 5-(cyanomethyl)furan-2-carboxylate

Precaución: for Research Use Only

Formula: C₉H₉NO₃
MW: 179.173 g/mol
MDL Number: MFCD12403566
CAS Number: 51129-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR