4-Bromo-2,3-dichlorobenzoic acid 95%

Supplier: Ambeed
Warning

A558412-5G A558412-1G
77103-586EA 625.94 USD
77103-586 77686-435
4-Bromo-2,3-dichlorobenzoic acid 95%
4-Bromo-2,3-dichlorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrCl₂O₂
MW: 269.90 g/mol
MDL Number: MFCD19686181
CAS Number: 1257517-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR