5-Ethynylbenzene-1,3-dioic acid 95%

Supplier: Ambeed
Warning

Synonyms: 5-Ethynylisophthalic acid

A558087-100MG A558087-1G A558087-250MG A558087-5G
77103-578EA 45.62 USD
77103-578 77103-580 77103-582 77103-584
5-Ethynylbenzene-1,3-dioic acid 95%
5-Ethynylbenzene-1,3-dioic acid

Precaución: for Research Use Only

Formula: C₁₀H₆O₄
MW: 190.152 g/mol
MDL Number: MFCD14635840
CAS Number: 432025-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR