Furan-2,4-dicarboxylic acid 95%

Supplier: Ambeed
Warning

A555964-100MG A555964-1G A555964-250MG
77103-570EA 108 USD
77103-570 77103-572 77103-574
Furan-2,4-dicarboxylic acid 95%
Formula: C₆H₄O₅
MDL Number: MFCD00159521
CAS Number: 4282-28-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR