(3-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A554840-1G A554840-250MG A554840-5G
77103-554EA 218 USD
77103-554 77103-556 77103-558
(3-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid 95%
(3-((1-(tert-Butoxycarbonyl)piperidin-4-yl)oxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₄BNO₅
MDL Number: MFCD18383744
CAS Number: 1224449-32-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR