1-Bromo-3,5-dinitrobenzene 98%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dinitrobromobenzene

A554548-250MG A554548-5G A554548-1G
77103-552EA 6.37 USD
77103-552 77551-672 77572-752
1-Bromo-3,5-dinitrobenzene 98%
1-Bromo-3,5-dinitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃BrN₂O₄
MW: 247 g/mol
MDL Number: MFCD00156596
CAS Number: 18242-39-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR