3-Cyclohexyl-D-alanine hydrate 98+%

Supplier: Ambeed
Warning

A554337-1G A554337-25G A554337-5G
77103-546EA 9 USD
77103-546 77103-548 77103-550
3-Cyclohexyl-D-alanine hydrate 98+%
3-Cyclohexyl-D-alanine hydrate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MDL Number: MFCD00217056
CAS Number: 213178-94-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR