4-Chloro-6-methoxypyrido[3,2-d]pyrimidine 97%

Supplier: Ambeed
Warning

A553125-100MG A553125-250MG
77103-534EA 81.69 USD
77103-534 77103-536
4-Chloro-6-methoxypyrido[3,2-d]pyrimidine 97%
4-Chloro-6-methoxypyrido[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₈H₆ClN₃O
MW: 195.60 g/mol
MDL Number: MFCD20258134
CAS Number: 1417555-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR