Ethyl piperazin-1-ylacetate 98%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-(piperazin-1-yl)acetate

A553079-5G A553079-25G
77103-532EA 23.34 USD
77103-532 77686-421
Ethyl piperazin-1-ylacetate 98%
Ethyl piperazin-1-ylacetate

Precaución: for Research Use Only

Formula: C₈H₁₆N₂O₂
MW: 172.23 g/mol
MDL Number: MFCD00040739
CAS Number: 40004-08-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR