5-Bromonicotinamide 97%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-pyridinecarboxamide

A552326-1G A552326-25G A552326-10G A552326-5G
77103-528EA 6.37 USD
77103-528 77103-530 77686-419 77686-420
5-Bromonicotinamide 97%
5-Bromonicotinamide

Precaución: for Research Use Only

Formula: C₆H₅BrN₂O
MW: 201.03 g/mol
MDL Number: MFCD00173919
CAS Number: 28733-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR