N-(3,4-Dimethoxyphenyl)-6,7-dimethoxyquinazolin-4-amine 98%

Supplier: Ambeed
Warning

A552084-100MG A552084-1G A552084-250MG
77103-522EA 153.84 USD
77103-522 77103-524 77103-526
N-(3,4-Dimethoxyphenyl)-6,7-dimethoxyquinazolin-4-amine 98%
N-(3,4-Dimethoxyphenyl)-6,7-dimethoxyquinazolin-4-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₉N₃O₄
MW: 341.36 g/mol
MDL Number: MFCD08147508
CAS Number: 202475-65-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR