10-Undecynoic acid 98%

Supplier: Ambeed
Warning

Synonyms: Undec-10-ynoic acid

A550617-100MG A550617-1G A550617-250MG A550617-25G A550617-5G
77103-512EA 9.55 USD
77103-512 77103-514 77103-516 77103-518 77103-520
10-Undecynoic acid 98%
10-Undecynoic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₈O₂
MW: 182.26 g/mol
MDL Number: MFCD00014389
CAS Number: 2777-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR