6-Bromo-2-methoxypyridine-3-boronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-2-methoxypyridin-3-ylboronic acid

A549867-100MG A549867-1G A549867-250MG
77103-502EA 21.22 USD
77103-502 77103-504 77103-506
6-Bromo-2-methoxypyridine-3-boronic acid 98%
6-Bromo-2-methoxypyridine-3-boronic acid

Precaución: for Research Use Only

Formula: C₆H₇BBrNO₃
MW: 231.83 g/mol
MDL Number: MFCD18382838
CAS Number: 1242314-44-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR