Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: Fmoc-4-[2-(Boc-amino)ethoxy]-L-phenylalanine

A549314-100G A549314-1G A549314-5G A549314-25G
77103-492EA 833.87 USD
77103-492 77103-494 77551-660 77594-678
Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine 97%
Fmoc-4-[2-(boc-amino)ethoxy]-L-phenylalanine

Precaución: for Research Use Only

Formula: C₃₁H₃₄N₂O₇
MW: 546.63 g/mol
MDL Number: MFCD07781260
CAS Number: 1013883-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR