7-Bromoisoquinoline-1,3(2H,4H)-dione 95%

Supplier: Ambeed
Warning

A548314-100MG A548314-1G A548314-250MG
77103-486EA 49 USD
77103-486 77103-488 77103-490
7-Bromoisoquinoline-1,3(2H,4H)-dione 95%
7-Bromoisoquinoline-1,3(2H,4H)-dione

Precaución: for Research Use Only

Formula: C₉H₆BrNO₂
MDL Number: MFCD12756049
CAS Number: 1033330-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR