(4-(Di-p-tolylamino)phenyl)boronic acid 95%

Supplier: Ambeed
Warning

A547709-250MG A547709-5G A547709-1G
77103-478EA 15 USD
77103-478 77103-480 77686-407
(4-(Di-p-tolylamino)phenyl)boronic acid 95%
(4-(Di-p-tolylamino)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₂₀H₂₀BNO₂
MDL Number: MFCD11977300
CAS Number: 654067-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR