(-)-Gallocatechin gallate 98%

Supplier: Ambeed
Warning

Synonyms: (2S,3R)-5,7-Dihydroxy-2-(3,4,5-trihydroxyphenyl)chroman-3-yl 3,4,5-trihydroxybenzoate

A547196-100MG A547196-50MG
77103-474EA 282 USD
77103-474 77103-476
(-)-Gallocatechin gallate 98%
(-)-Gallocatechin gallate

Precaución: for Research Use Only

Formula: C₂₂H₁₈O₁₁
MW: 458.38 g/mol
MDL Number: MFCD00214298
CAS Number: 4233-96-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR