2-Amino-6-benzyloxypyridine 98%

Supplier: Ambeed
Warning

A546741-1G A546741-250MG
77103-472EA 166 USD
77103-472 77900-505
2-Amino-6-benzyloxypyridine 98%
2-Amino-6-benzyloxypyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MDL Number: MFCD21333044
CAS Number: 1174626-28-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR