Cyclopropanemethylamine 95%

Supplier: Ambeed
Danger

A544145-10G A544145-1G A544145-25G A544145-5G
77103-450EA 38.2 USD
77103-450 77103-452 77103-454 77103-456
Cyclopropanemethylamine 95%
Cyclopropanemethylamine

Precaución: for Research Use Only

Formula: C₄H₉N
MW: 71.12 g/mol
MDL Number: MFCD00037147
CAS Number: 2516-47-4
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR