2-Fluorobenzoyl chloride 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
o-Fluorobenzoyl chloride
A542426-25G
A542426-100G
77103-442EA
16
USD
77103-442
77686-394
2-Fluorobenzoyl chloride 95%
2-Fluorobenzoyl chloride
Precaución: for Research Use Only
Formula:
C₇H₄ClFO MW: 158.56 g/mol Boiling Pt: 206 |
MDL Number:
MFCD00000656 CAS Number: 393-52-2 UN: 3265 ADR: 8 Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...