2-Mercapto-5-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Methylpyridine-2(1H)-thione

A540044-1G A540044-250MG
77103-436EA 97 USD
77103-436 77103-438
2-Mercapto-5-methylpyridine 97%
2-Mercapto-5-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇NS
MW: 125.191 g/mol
MDL Number: MFCD02684159
CAS Number: 18368-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR