3-(4-Acetylphenyl)-2-aminopropanoic acid hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A537966-100MG
A537966-1MG
77103-416EA
29.71
USD
77103-416
77686-386
3-(4-Acetylphenyl)-2-aminopropanoic acid hydrochloride 95%
3-(4-Acetylphenyl)-2-aminopropanoic acid hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₄ClNO₃ MW: 243.69 g/mol |
MDL Number:
MFCD08436130 CAS Number: 1360436-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...