3-Butylthiophene 98%
Supplier: Ambeed
|
Danger
|
A537541-10G
A537541-1G
A537541-5G
A537541-25G
77103-410EA
67.9
USD
77103-410
77103-412
77103-414
77686-385
3-Butylthiophene 98%
3-Butylthiophene
Precaución: for Research Use Only
Formula:
C₈H₁₂S MW: 140.2459 g/mol |
MDL Number:
MFCD00143179 CAS Number: 34722-01-5 UN: 1993 ADR: 3,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...