3-Nitroaniline hydrochloride 98%
Supplier: Ambeed
|
Warning
|
A536981-10G
A536981-25G
A536981-5G
77103-396EA
29
USD
77103-396
77103-398
77103-400
3-Nitroaniline hydrochloride 98%
3-Nitroaniline hydrochloride
Precaución: for Research Use Only
Formula:
C₆H₇ClN₂O₂ |
MDL Number:
MFCD00050693 CAS Number: 33240-96-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...