4-Methyl-7-nitro-1H-indole-3-carbonitrile 95%

Supplier: Ambeed
Warning

A536360-100MG A536360-1G A536360-5G
77103-378EA 42 USD
77103-378 77103-380 77103-382
4-Methyl-7-nitro-1H-indole-3-carbonitrile 95%
4-Methyl-7-nitro-1H-indole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₀H₇N₃O₂
MDL Number: MFCD17779298
CAS Number: 289483-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR