4-(2-Methoxyethoxy)benzenesulfonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 4-(2-Methoxyethoxy)-phenylsulphonyl chloride

A532989-100MG A532989-1G A532989-250MG
77103-348EA 47.75 USD
77103-348 77103-350 77103-352
4-(2-Methoxyethoxy)benzenesulfonyl chloride 95%
4-(2-Methoxyethoxy)benzenesulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₁₁ClO₄S
MW: 250.699 g/mol
MDL Number: MFCD09816161
CAS Number: 204072-53-7
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR