(S)-3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid 95%

Supplier: Ambeed
Warning

A532931-100MG A532931-250MG A532931-1G A532931-5G
77103-344EA 15 USD
77103-344 77103-346 77613-678 77901-130
(S)-3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid 95%
(S)-3-((tert-Butoxycarbonyl)amino)-2-methylpropanoic acid

Precaución: for Research Use Only

Formula: C₉H₁₇NO₄
MDL Number: MFCD04040045
CAS Number: 190897-47-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR