Ethyl-4-methyloxazole-5-carboxylate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 4-methyloxazole-5-carboxylate
A532843-100MG
A532843-1G
A532843-250MG
A532843-25G
A532843-5G
A532843-10G
77103-334EA
9.55
USD
77103-334
77103-336
77103-338
77103-340
77103-342
77594-644
Ethyl-4-methyloxazole-5-carboxylate 97%
Ethyl-4-methyloxazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₉NO₃ MW: 155.15 g/mol |
MDL Number:
MFCD00062573 CAS Number: 20485-39-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...