2-Amino-3,4-difluorobenzonitrile 99%

Supplier: Ambeed
Danger

A532807-100MG A532807-1G A532807-250MG A532807-5G
77103-326EA 8 USD
77103-326 77103-328 77103-330 77103-332
2-Amino-3,4-difluorobenzonitrile 99%
2-Amino-3,4-difluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄F₂N₂
MDL Number: MFCD21606584
CAS Number: 1384265-40-0
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR