6-Bromo-3-chloro-2-fluoropyridine 98%

Supplier: Ambeed
Warning

A529844-100MG A529844-1G A529844-250MG
77103-306EA 26 USD
77103-306 77103-308 77103-310
6-Bromo-3-chloro-2-fluoropyridine 98%
6-Bromo-3-chloro-2-fluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂BrClFN
MDL Number: MFCD18257574
CAS Number: 1211578-30-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR