Cyclobutylamine 98%

Supplier: Ambeed
Danger

Synonyms: Cyclobutylamine, Cyclobutanamine, Aminocyclobutane

A529599-5G A529599-1G A529599-25G
77103-304EA 57.29 USD
77103-304 77551-598 77686-363
Cyclobutylamine 98%
Cyclobutylamine

Precaución: for Research Use Only

Formula: C₄H₉N
MW: 71.12 g/mol
MDL Number: MFCD00001328
CAS Number: 2516-34-9
UN: 2733
ADR: 3,8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR