Methyl 4-ethanesulfonamidobenzoate 97%

Supplier: Ambeed
Warning

A529093-10G A529093-1G
77103-298EA 150 USD
77103-298 77103-300
Methyl 4-ethanesulfonamidobenzoate 97%
Methyl 4-ethanesulfonamidobenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₄S
MDL Number: MFCD03379139
CAS Number: 544451-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR