3-Bromo-4,5-dichlorotoluene 95%

Supplier: Ambeed
Warning

A528826-1G A528826-5G
77103-294EA 21.99 USD
77103-294 77103-296
3-Bromo-4,5-dichlorotoluene 95%
3-Bromo-4,5-dichlorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrCl₂
MDL Number: MFCD26792624
CAS Number: 960305-14-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR