(E)-3-(Furan-3-yl)acrylic acid 98%

Supplier: Ambeed
Warning

A528636-1G A528636-250MG A528636-5G
77103-288EA 50.93 USD
77103-288 77103-290 77103-292
(E)-3-(Furan-3-yl)acrylic acid 98%
(E)-3-(Furan-3-yl)acrylic acid

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
MDL Number: MFCD00075074
CAS Number: 81311-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR