OF-1 98+%

Supplier: Ambeed
Danger

Synonyms: 4-Bromo-N-(6-methoxy-1,3-dimethyl-2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-methylbenzenesulphonamide

A525019-10MG A525019-1MG A525019-50MG A525019-5MG
77103-254EA 91.24 USD
77103-254 77103-256 77103-258 77103-260
OF-1 98+%
OF-1

Precaución: for Research Use Only

Formula: C₁₇H₁₈BrN₃O₄S
MW: 440.31 g/mol
MDL Number: MFCD26964046
CAS Number: 919973-83-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR