2-(Dimethoxyphosphoryl)acetic acid 95%

Supplier: Ambeed
Warning

A524106-100MG A524106-1G A524106-250MG
77103-248EA 25 USD
77103-248 77103-250 77901-157
2-(Dimethoxyphosphoryl)acetic acid 95%
2-(Dimethoxyphosphoryl)acetic acid

Precaución: for Research Use Only

Formula: C₄H₉O₅P
MDL Number: MFCD03788193
CAS Number: 34159-46-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR