2-Bromo-6-isopropoxynaphthalene 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-6-(propan-2-yloxy)naphthalene

A523900-1G A523900-250MG A523900-5G
77103-242EA 76.39 USD
77103-242 77103-244 77103-246
2-Bromo-6-isopropoxynaphthalene 98%
2-Bromo-6-isopropoxynaphthalene

Precaución: for Research Use Only

Formula: C₁₃H₁₃BrO
MW: 265.15 g/mol
MDL Number: MFCD18262542
CAS Number: 200875-37-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR