N-Benzyl-5-fluoro-2-nitroaniline 95%

Supplier: Ambeed
Warning

A522801-1G A522801-5G
77103-232EA 11 USD
77103-232 77103-234
N-Benzyl-5-fluoro-2-nitroaniline 95%
N-Benzyl-5-fluoro-2-nitroaniline

Precaución: for Research Use Only

Formula: C₁₃H₁₁FN₂O₂
MW: 246.23 g/mol
MDL Number: MFCD17336280
CAS Number: 131885-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR