8-Bromo-1-chlorodibenzo[b,d]furan 97%

Supplier: Ambeed
Warning

A519790-1G A519790-5G
77103-212EA 23.34 USD
77103-212 77103-214
8-Bromo-1-chlorodibenzo[b,d]furan 97%
8-Bromo-1-chlorodibenzo[b,d]furan

Precaución: for Research Use Only

Formula: C₁₂H₆BrClO
MW: 281.53 g/mol
CAS Number: 2225909-61-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR