Methyl 3-bromo-4-fluoro-5-nitrobenzoate 98%

Supplier: Ambeed
Warning

A519366-1G A519366-250MG A519366-5G
77103-206EA 53 USD
77103-206 77103-208 77103-210
Methyl 3-bromo-4-fluoro-5-nitrobenzoate 98%
Methyl 3-bromo-4-fluoro-5-nitrobenzoate

Precaución: for Research Use Only

Formula: C₈H₅BrFNO₄
MDL Number: MFCD22549345
CAS Number: 1403483-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR