2-Cyano-3-phenyl-acrylic acid methyl ester 95%

Supplier: Ambeed
Warning

A518785-1G A518785-5G
77103-202EA 65 USD
77103-202 77103-204
2-Cyano-3-phenyl-acrylic acid methyl ester 95%
2-Cyano-3-phenyl-acrylic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₁H₉NO₂
MDL Number: MFCD00025864
CAS Number: 14533-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR